missing translation for 'onlineSavingsMsg'
Learn More

3-(Trimethylsilyl)propargyl bromide, 97%

Product Code. 10441294 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10441294 1 g 1g
10091364 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10441294 Supplier Thermo Scientific Acros Supplier No. 399840010
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 38002-45-8 | C6H11BrSi | 191.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38002-45-8
Molecular Formula C6H11BrSi
Molecular Weight (g/mol) 191.14
MDL Number MFCD00134460
InChI Key GAPRPFRDVCCCHR-UHFFFAOYSA-N
Synonym 3-bromo-1-trimethylsilyl-1-propyne, 3-bromoprop-1-yn-1-yl trimethylsilane, 3-bromo-1-propynyl trimethylsilane, 3-trimethylsilyl propargyl bromide, 3-bromoprop-1-ynyl trimethyl silane, silane, 3-bromo-1-propynyl trimethyl, 3-bromoprop-1-yn-1-yl trimethyl silane, acmc-1cthe, ksc496i3r
PubChem CID 642589
IUPAC Name 3-bromoprop-1-ynyl(trimethyl)silane
SMILES C[Si](C)(C)C#CCBr

Specifications

Density 1.3510g/mL
Boiling Point 44°C to 45°C (2.0 mmHg)
Flash Point 62°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3)3SiC∽CCH2Br
Quantity 1 g
Specific Gravity 1.351
Solubility Information Solubility in water: insoluble
Formula Weight 191.14
Percent Purity 97%
Chemical Name or Material 3-(Trimethylsilyl)propargyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 3-(Trimethylsilyl)propargyl bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.