accessibility menu, dialog, popup

UserName

H3CH3CCH3OONHBOHHO

CAS RN 380430-49-9

IUPAC Name: (4-{[(tert-butoxy)carbonyl]amino}phenyl)boronic acid
Synonyms: 4-[(tert-butoxycarbonyl)amino]phenylboronic acid
Molecular Weight (g/mol): 237.06
Molecular Formula: C11H16BNO4
InChi Key: UBVOLHQIEQVXGM-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC=C(C=C1)B(O)O
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)