accessibility menu, dialog, popup

UserName

CAS RN 38460-95-6

ClOCH2

CAS RN 38460-95-6

IUPAC Name: undec-10-enoyl chloride
Synonyms: 10-undecenoyl chloride
Molecular Weight (g/mol): 202.72
Molecular Formula: C11H19ClO
InChi Key: MZFGYVZYLMNXGL-UHFFFAOYSA-N
SMILES: ClC(=O)CCCCCCCCC=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)