missing translation for 'onlineSavingsMsg'
Learn More

10-Undecenoyl chloride, 98%

Product Code. 10387970 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 10387970

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38460-95-6 | C11H19ClO | 202.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38460-95-6
Molecular Formula C11H19ClO
Molecular Weight (g/mol) 202.72
MDL Number MFCD00000772
InChI Key MZFGYVZYLMNXGL-UHFFFAOYSA-N
Synonym 10-undecenoyl chloride, 10-undecylenoyl chloride, 10-undecenoylchloride, omega-undecylenic acid chloride, undecylenoyl chloride, .omega.-undecylenic acid chloride, acmc-1ckgs, 10-hendecenoyl chloride, 10-undecenoic acid chloride, 4-02-00-01615 beilstein handbook reference
PubChem CID 38042
IUPAC Name undec-10-enoyl chloride
SMILES ClC(=O)CCCCCCCCC=C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 0.9400g/mL
Boiling Point 120°C to 122°C (10.0 mmHg)
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2C=CH(CH2)8COCl
Refractive Index 1.4532 to 1.4552
Quantity 500 mL
Beilstein 02, 459
Specific Gravity 0.94
Formula Weight 202.72
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 10-Undecenoyl chloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 10-Undecenoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.