CAS RN 385-01-3
CAS RN 385-01-3
IUPAC Name:
3-fluoro-2-nitrophenol
Synonyms:
3-fluoro-2-nitrophenol
Molecular Weight (g/mol):
157.1
Molecular Formula:
C6H4FNO3
InChi Key:
GAWNBKUTBVLIPL-UHFFFAOYSA-N
SMILES:
OC1=C(C(F)=CC=C1)[N+]([O-])=O
1
–
1
of
1
results
1
3-Fluoro-2-nitrophenol, 97%
CAS: 385-01-3 Molecular Formula: C6H4FNO3 Molecular Weight (g/mol): 157.1 MDL Number: MFCD07368753 InChI Key: GAWNBKUTBVLIPL-UHFFFAOYSA-N Synonym: 3-fluoro-2-nitro-phenol,2-nitro-3-fluorophenol,phenol, 3-fluoro-2-nitro,3-fluoronitro-phenol,pubchem4124,acmc-209j0i,ksc497m1h PubChem CID: 2782777 IUPAC Name: 3-fluoro-2-nitrophenol SMILES: C1=CC(=C(C(=C1)F)[N+](=O)[O-])O