accessibility menu, dialog, popup

UserName

CAS RN 38506-97-7

H3CCH3HNO

CAS RN 38506-97-7

IUPAC Name: 1-phenyl-2-[(propan-2-yl)amino]ethan-1-one
Synonyms: 2-(isopropylamino)-1-phenylethanone
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: IHVMTNLVDVWAAA-UHFFFAOYSA-N
SMILES: CC(C)NCC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results