missing translation for 'onlineSavingsMsg'
Learn More

2-(Isopropylamino)acetophenone hydrochloride, 98%, Thermo Scientific™

Product Code. 11484780
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
11484780 1 g 1g
11474780 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 11484780

Brand: Thermo Scientific Alfa Aesar H50799.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38506-97-7
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD18785640
InChI Key IHVMTNLVDVWAAA-UHFFFAOYSA-N
Synonym 2-isopropylamino-1-phenylethanone, 2-isopropylamino acetophenone, phenyl isopropylaminomethyl ketone, 2-methylethyl amino-1-phenylethan-1-one, 1-phenyl-2-propan-2-yl amino ethan-1-one, 2-isopropylamino acetophenone hydrochloride
PubChem CID 12861561
IUPAC Name 1-phenyl-2-(propan-2-ylamino)ethanone
SMILES CC(C)NCC(=O)C1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 213.71
Percent Purity 98%
Chemical Name or Material 2-(Isopropylamino)acetophenone hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.