accessibility menu, dialog, popup

UserName

HOOOO

CAS RN 38559-92-1

IUPAC Name: 2-[4-(benzyloxy)phenoxy]acetic acid
Synonyms: 4-(benzyloxy)phenoxyacetic acid
Molecular Weight (g/mol): 258.27
Molecular Formula: C15H14O4
InChi Key: VXMSXBVTUNOSLL-UHFFFAOYSA-N
SMILES: OC(=O)COC1=CC=C(OCC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results