accessibility menu, dialog, popup

UserName

ON

CAS RN 38707-70-9

IUPAC Name: quinoline-8-carbaldehyde
Synonyms: quinoline-8-carbaldehyde
Molecular Weight (g/mol): 157.17
Molecular Formula: C10H7NO
InChi Key: OVZQVGZERAFSPI-UHFFFAOYSA-N
SMILES: O=CC1=C2N=CC=CC2=CC=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)