missing translation for 'onlineSavingsMsg'
Learn More

8-Quinolinecarbaldehyde, Thermo Scientific™

Product Code. 10610903
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10610903 1 g 1g
10461494 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10610903 Supplier Thermo Scientific Maybridge Supplier No. CC04304DA
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 38707-70-9
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.172
MDL Number MFCD00805837
InChI Key OVZQVGZERAFSPI-UHFFFAOYSA-N
Synonym 8-quinolinecarboxaldehyde, 8-quinolinecarbaldehyde, quinoline-8-carboxaldehyde, 8-quinoline carbaldehyde, 8-formylquinoline, ccris 6968, timtec-bb sbb010129, 8-quinolinecarboaldehyde, acmc-209j1k, ksc224g8j
PubChem CID 170103
IUPAC Name quinoline-8-carbaldehyde
SMILES C1=CC2=C(C(=C1)C=O)N=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 157.17
Percent Purity 97%
Chemical Name or Material 8-Quinolinecarbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.