accessibility menu, dialog, popup

UserName

SP

CAS RN 3878-45-3

IUPAC Name: triphenyl-λ⁵-phosphanethione
Synonyms: triphenylphosphine sulfide
Molecular Weight (g/mol): 294.35
Molecular Formula: C18H15PS
InChi Key: VYNGFCUGSYEOOZ-UHFFFAOYSA-N
SMILES: S=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results