accessibility menu, dialog, popup

UserName

HOOHHO

CAS RN 3897-89-0

IUPAC Name: 4-(hydroxymethyl)benzene-1,2-diol
Synonyms: 3,4-dihydroxybenzyl alcohol
Molecular Weight (g/mol): 140.14
Molecular Formula: C7H8O3
InChi Key: PCYGLFXKCBFGPC-UHFFFAOYSA-N
SMILES: OCC1=CC=C(O)C(O)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results