accessibility menu, dialog, popup

UserName

CH3OOFOH

CAS RN 392-04-1

IUPAC Name: methyl 4-fluoro-2-hydroxybenzoate
Synonyms: methyl 4-fluoro-2-hydroxybenzoate
Molecular Weight (g/mol): 170.14
Molecular Formula: C8H7FO3
InChi Key: WPWUDDDJTIZBGL-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(O)C=C(F)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results