accessibility menu, dialog, popup

UserName

CAS RN 39230-37-0

NaOO2H2H2H

CAS RN 39230-37-0

IUPAC Name: sodium (²H₃)acetate
Synonyms: sodium (²H₃)acetate
Molecular Weight (g/mol): 85.05
Molecular Formula: C2H3NaO2
InChi Key: VMHLLURERBWHNL-NIIDSAIPSA-M
SMILES: [Na+].[2H]C([2H])([2H])C([O-])=O
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results