accessibility menu, dialog, popup

UserName

NH2FFFOHO

CAS RN 393-06-6

IUPAC Name: 4-amino-2-(trifluoromethyl)benzoic acid
Synonyms: 4-amino-2-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 205.14
Molecular Formula: C8H6F3NO2
InChi Key: AMVHEVZYTGHASE-UHFFFAOYSA-N
SMILES: NC1=CC(=C(C=C1)C(O)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results