accessibility menu, dialog, popup

UserName

CAS RN 393-52-2

FClO

CAS RN 393-52-2

IUPAC Name: 2-fluorobenzoyl chloride
Synonyms: benzoyl chloride, 2-fluoro-
Molecular Weight (g/mol): 158.56
Molecular Formula: C7H4ClFO
InChi Key: RAAGZOYMEQDCTD-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1C(Cl)=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)