CAS RN 3939-14-8
CAS RN 3939-14-8
IUPAC Name:
2-fluoropyridine-4-carbonitrile
Synonyms:
2-fluoropyridine-4-carbonitrile
Molecular Weight (g/mol):
122.1
Molecular Formula:
C6H3FN2
InChi Key:
DYQMGPSBUBTNJZ-UHFFFAOYSA-N
SMILES:
FC1=CC(=CC=N1)C#N
1
–
1
of
1
results
1
4-Cyano-2-fluoropyridine, 97%
CAS: 3939-14-8 Molecular Formula: C6H3FN2 Molecular Weight (g/mol): 122.102 MDL Number: MFCD09027299 InChI Key: DYQMGPSBUBTNJZ-UHFFFAOYSA-N PubChem CID: 12002587 IUPAC Name: 2-fluoropyridine-4-carbonitrile SMILES: C1=CN=C(C=C1C#N)F