CAS RN 394-30-9
CAS RN 394-30-9
IUPAC Name:
5-chloro-2-fluorobenzoate
Synonyms:
5-chloro-2-fluorobenzoate
Molecular Weight (g/mol):
173.55
Molecular Formula:
C7H3ClFO2
InChi Key:
WGAVMKXCDMQVNF-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC(Cl)=CC=C1F
1
–
2
of
2
results
2
Alfa Aesar™ 5-Chloro-2-fluorobenzoic acid, 97%
CAS: 394-30-9 Molecular Formula: C7H4ClFO2 Molecular Weight (g/mol): 174.555 MDL Number: MFCD00665762 InChI Key: WGAVMKXCDMQVNF-UHFFFAOYSA-N Synonym: 2-fluoro-5-chlorobenzoic acid,5-chloro-2-fluoro-benzoic acid,5-chloro-2-fluorobenzoicacid,benzoic acid, 5-chloro-2-fluoro,pubchem1375,acmc-1aigz,ksc223m7h,rarechem al bo 0944,5-chloro-2-fluorobenzoic acid,benzoicacid, 5-chloro-2-fluoro PubChem CID: 2736537 IUPAC Name: 5-chloro-2-fluorobenzoic acid SMILES: C1=CC(=C(C=C1Cl)C(=O)O)F