accessibility menu, dialog, popup

UserName

FH2C

CAS RN 394-46-7

IUPAC Name: 1-ethenyl-2-fluorobenzene
Synonyms: 1-ethenyl-2-fluorobenzene
Molecular Weight (g/mol): 122.14
Molecular Formula: C8H7F
InChi Key: YNQXOOPPJWSXMW-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1C=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results