CAS RN 39450-01-6
CAS RN 39450-01-6
IUPAC Name:
{[6-(1-hydroxyethyl)-4-methyl-2-{[(4-nitrophenyl)methoxy]carbonyl}-7-oxo-3,4-diphenoxy-1-azabicyclo[3.2.0]hept-2-en-5-yl]oxy}phosphonic acid
Synonyms:
[6-(1-hydroxyethyl)-4-methyl-2-{[(4-nitrophenyl)methoxy]carbonyl}-7-oxo-3,4-diphenoxy-1-azabicyclo[3.2.0]hept-2-en-5-yl]oxyphosphonic acid
Molecular Weight (g/mol):
626.51
Molecular Formula:
C29H27N2O12P
InChi Key:
OUPAKHMWUJJXOM-UHFFFAOYNA-N
SMILES:
CC(O)C1C(=O)N2C(C(=O)OCC3=CC=C(C=C3)[N+]([O-])=O)=C(OC3=CC=CC=C3)C(C)(OC3=CC=CC=C3)C12OP(O)(O)=O
1
–
2
of
2
results
1
Melford Proteinase K
From Tritirachium album.
2
Thermo Scientific™ Proteinase K, Ready-to-Use Soln.
A stable and highly reactive serine protease; used for protein and nucleic acid isolation