accessibility menu, dialog, popup

UserName

CAS RN 395-23-3

IUPAC Name: phenyl[4-(trifluoromethyl)phenyl]methanol
Synonyms: phenyl[4-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 252.24
Molecular Formula: C14H11F3O
InChi Key: LIZDGCXCDJOWBS-UHFFFAOYNA-N
SMILES: OC(C1=CC=CC=C1)C1=CC=C(C=C1)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results