accessibility menu, dialog, popup

UserName

CH3OH2NNH2

CAS RN 39549-79-6

IUPAC Name: 2-amino-4-methylbenzamide
Synonyms: 2-amino-4-methylbenzamide
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: RUHKZVAPXHIWJH-UHFFFAOYSA-N
SMILES: CC1=CC(N)=C(C=C1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results