accessibility menu, dialog, popup

UserName

HOOSN

CAS RN 3973-08-8

IUPAC Name: 1,3-thiazole-4-carboxylic acid
Synonyms: 1,3-thiazole-4-carboxylic acid
Molecular Weight (g/mol): 129.13
Molecular Formula: C4H3NO2S
InChi Key: HMVYYTRDXNKRBQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CSC=N1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results