missing translation for 'onlineSavingsMsg'
Learn More

4-Thiazolecarboxylic acid, 97%

Product Code. 15562293 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15562293 5 g 5g
15552293 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15562293 Supplier Thermo Scientific Acros Supplier No. 443970050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3973-08-8 | C4H3NO2S | 129.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3973-08-8
Molecular Formula C4H3NO2S
Molecular Weight (g/mol) 129.14

Specifications

Melting Point 195.0°C to 199.0°C
Loss on Drying 3% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Synonym 1,1,4,4-tetraethoxy-2-butyne, 2-butyne,1,1,4,4-tetraethoxy
InChI Key QJVVNQLFPMBGLC-UHFFFAOYSA-N
SMILES CCOC(C#CC(OCC)OCC)OCC
IUPAC Name 1,1,4,4-tetraethoxybut-2-yne
PubChem CID 2724486
Formula Weight 129.14
Percent Purity 97%
Chemical Name or Material 4-Thiazolecarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Thiazolecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.