accessibility menu, dialog, popup

UserName

CAS RN 39739-60-1

CH3CH3HNOH3CHCl

CAS RN 39739-60-1

IUPAC Name: methyl 2-methylpropanimidate hydrochloride
Synonyms: methyl 2-methylpropanimidate hydrochloride
Molecular Weight (g/mol): 137.61
Molecular Formula: C5H12ClNO
InChi Key: RMXGPHAHPIIESQ-UHFFFAOYSA-N
SMILES: Cl.COC(=N)C(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)