missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-methylpropionimidate hydrochloride, 97%

Product Code. 15420059
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420059 5 g 5g
15448286 250 mg 250mg
15410059 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15420059 Supplier Thermo Scientific Alfa Aesar Supplier No. H63386.06
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 39739-60-1 | C5H12ClNO | 137.61 g/mol

Methyl 2-methylpropionimidate hydrochloride, is used as a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-methylpropionimidate hydrochloride, is used as a fine chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39739-60-1
Molecular Formula C5H12ClNO
Molecular Weight (g/mol) 137.61
MDL Number MFCD03844704
InChI Key RMXGPHAHPIIESQ-UHFFFAOYSA-N
Synonym methyl 2-methylpropanecarboximidate hydrochloride, methyl 2-methylpropanimidate hydrochloride, methyl 2-methylpropanimidic acid hydrochloride, methyl 2-methylpropanimidoate hydrochloride, methyl isobutyrimidate hydrochloride, methyl 2-methylpropanimidate-hydrogen chloride 1/1, methyl 1z-2-methylpropanimidoate hydrochloride 1:1
PubChem CID 371709
IUPAC Name methyl 2-methylpropanimidate;hydrochloride
SMILES Cl.COC(=N)C(C)C

Specifications

Color White
Linear Formula (CH3)2CHC(=NH)OCH3·HCl
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 137.61
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 2-methylpropionimidate hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.