accessibility menu, dialog, popup

UserName

CAS RN 39755-95-8

CH3OHNOO

CAS RN 39755-95-8

IUPAC Name: 5-methoxy-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 5-methoxy-1H-indole-2,3-dione
Molecular Weight (g/mol): 177.16
Molecular Formula: C9H7NO3
InChi Key: DMHGXMPXHPOXBF-UHFFFAOYSA-N
SMILES: COC1=CC=C2NC(=O)C(=O)C2=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)