missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyisatin, 97%

Product Code. 13565030 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13565030 100 g 100g
13545030 5 g 5g
13555030 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13565030 Supplier Thermo Scientific Acros Supplier No. 448831000
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
This item is not returnable. View return policy

CAS: 39755-95-8 | C9H7NO3 | 177.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39755-95-8
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.16
MDL Number MFCD00169023
InChI Key DMHGXMPXHPOXBF-UHFFFAOYSA-N
Synonym 5-methoxyisatin, 5-methoxyindoline-2,3-dione, 5-methoxy-2,3-indolinedione, 1h-indole-2,3-dione, 5-methoxy, 5-methoxy-indole-2,3-dione, 5-methoxy isatin, 5-methoxyindole-2,3-dione, 5-methoxy-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-methoxy, 1h-indole-2,3-dione, 5-methoxy-9ci
PubChem CID 38333
IUPAC Name 5-methoxy-1H-indole-2,3-dione
SMILES COC1=CC=C2NC(=O)C(=O)C2=C1

Specifications

Melting Point 201°C to 203°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 100 g
Formula Weight 177.16
Percent Purity 97%
Chemical Name or Material 5-Methoxyisatin
compliance-icons
Product Identifier
  • 5-Methoxyisatin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.