CAS RN 3982-91-0
CAS RN 3982-91-0
IUPAC Name:
phosphorothioyl trichloride
Synonyms:
thiophosphoryl chloride
Molecular Weight (g/mol):
169.38
Molecular Formula:
Cl3PS
InChi Key:
WQYSXVGEZYESBR-UHFFFAOYSA-N
SMILES:
ClP(Cl)(Cl)=S
1
–
1
of
1
results
1
Thiophosphoryl Chloride, TRC
CAS: 3982-91-0 Molecular Formula: Cl3PS Molecular Weight (g/mol): 169.4 Synonym: NSC 158334,Phosphorothionic Trichloride,Phosphorus Sulfide Trichloride IUPAC Name: trichloro(sulfanylidene)-λ^{5}-phosphane SMILES: ClP(=S)(Cl)Cl