missing translation for 'onlineSavingsMsg'
Learn More

Thiophosphoryl Chloride, TRC

Product Code. 30453614
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453614 100 g 100g
30452988 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30453614 Supplier Toronto Research Chemicals Supplier No. TRCT368203100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3982-91-0
Molecular Formula Cl3PS
Molecular Weight (g/mol) 169.4
Synonym NSC 158334, Phosphorothionic Trichloride, Phosphorus Sulfide Trichloride
IUPAC Name trichloro(sulfanylidene)-λ^{5}-phosphane
SMILES ClP(=S)(Cl)Cl

Specifications

Quantity 100 g
InChI Formula InChI=1S/Cl3PS/c1-4(2,3)5
Formula Weight 167.8524
Chemical Name or Material Thiophosphoryl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.