accessibility menu, dialog, popup

UserName

CAS RN 398456-87-6

IUPAC Name: 2-fluoro-4-methanesulfonylphenol
Synonyms: 2-fluoro-4-methanesulfonylphenol
Molecular Weight (g/mol): 190.19
Molecular Formula: C7H7FO3S
InChi Key: OVEMQCGVMRIIHW-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC(F)=C(O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results