accessibility menu, dialog, popup

UserName

ONOClFFF

CAS RN 39974-35-1

IUPAC Name: 2-chloro-1-nitro-3-(trifluoromethyl)benzene
Synonyms: 2-chloro-1-nitro-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 225.55
Molecular Formula: C7H3ClF3NO2
InChi Key: TWABRHQBRWLSSE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(=C1Cl)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results