accessibility menu, dialog, popup

UserName

CAS RN 39998-81-7

IUPAC Name: 2-fluoro-4-iodo-1-methylbenzene
Synonyms: 2-fluoro-4-iodo-1-methylbenzene
Molecular Weight (g/mol): 236.03
Molecular Formula: C7H6FI
InChi Key: QZLWTFTXGKKCHZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(I)C=C1F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results