accessibility menu, dialog, popup

UserName

H3COOFCl

CAS RN 401-56-9

IUPAC Name: ethyl 2-chloro-2-fluoroacetate
Synonyms: ethyl 2-chloro-2-fluoroacetate
Molecular Weight (g/mol): 140.54
Molecular Formula: C4H6ClFO2
InChi Key: WUHVJSONZHSDFC-UHFFFAOYNA-N
SMILES: CCOC(=O)C(F)Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results