missing translation for 'onlineSavingsMsg'
Learn More

Ethyl chlorofluoroacetate, 98%, Thermo Scientific™

Product Code. 11492267
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492267 5 g 5g
11402277 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11492267 Supplier Thermo Scientific Alfa Aesar Supplier No. A14196.06
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 401-56-9
Molecular Formula C4H6ClFO2
Molecular Weight (g/mol) 140.538
MDL Number MFCD00039333
InChI Key WUHVJSONZHSDFC-UHFFFAOYSA-N
Synonym ethyl chlorofluoroacetate, acetic acid, chlorofluoro-, ethyl ester, chlorofluoroacetic acid ethyl ester, ethyl chloro fluoro acetate, ethyl fluorochloroacetate, acetic acid, 2-chloro-2-fluoro-, ethyl ester, acmc-1csjg, ksc492m6l, ethyl 2-chloro-2-fluoro-acetate
PubChem CID 101668
IUPAC Name ethyl 2-chloro-2-fluoroacetate
SMILES CCOC(=O)C(F)Cl

Specifications

Density 1.225
Boiling Point 128°C to 130°C
Flash Point 44°C (111°F)
Odor Pungent
Refractive Index 1.396
Quantity 5 g
UN Number UN2920
Beilstein 1748679
Formula Weight 140.54
Percent Purity 98%
Chemical Name or Material Ethyl chlorofluoroacetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.