accessibility menu, dialog, popup

UserName

CAS RN 401892-81-7

SFFFFFOCl

CAS RN 401892-81-7

IUPAC Name: 3-(pentafluoro-λ⁶-sulfanyl)benzoyl chloride
Synonyms: 3-(pentafluoro-λ⁶-sulfanyl)benzoyl chloride
Molecular Weight (g/mol): 266.61
Molecular Formula: C7H4ClF5OS
InChi Key: IDHIADOQLRPGHK-UHFFFAOYSA-N
SMILES: FS(F)(F)(F)(F)C1=CC=CC(=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results