accessibility menu, dialog, popup

UserName

OOCl

CAS RN 40191-32-0

IUPAC Name: oxane-4-carbonyl chloride
Synonyms: oxane-4-carbonyl chloride
Molecular Weight (g/mol): 148.59
Molecular Formula: C6H9ClO2
InChi Key: RYGUCYSSMOFTSH-UHFFFAOYSA-N
SMILES: ClC(=O)C1CCOCC1

 No Refinements