missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-2H-pyran-4-carbonyl chloride, ≥97%, Thermo Scientific™

Product Code. 11365876
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365876 1 g 1g
10423682 250 mg 250mg
10332335 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11365876 Supplier Thermo Scientific Maybridge Supplier No. CC29902DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 40191-32-0
Molecular Formula C6H9ClO2
Molecular Weight (g/mol) 148.59
MDL Number MFCD06200863
InChI Key RYGUCYSSMOFTSH-UHFFFAOYSA-N
Synonym tetrahydro-2h-pyran-4-carbonyl chloride, 2h-pyran-4-carbonyl chloride, tetrahydro, tetrahydro-2h-pyran-4-carbonylchloride, pubchem12437, ksc235m5r, tetrahydropyran-4-carbonyl chloride, tetrahydro-pyran-4-carbonyl chloride, 4-chloroformyl tetrahydro-2h-pyran, tetrahydropyran-4-carboxylic chloride, 4-chlorocarbonyl tetrahydro-2h-pyran
PubChem CID 2795505
IUPAC Name oxane-4-carbonyl chloride
SMILES ClC(=O)C1CCOCC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 148.59
Percent Purity 95%
Chemical Name or Material Tetrahydro-2H-pyran-4-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.