CAS RN 4027-57-0
IUPAC Name:
ethyl 5-methyl-1H-pyrazole-3-carboxylate
Synonyms:
ethyl 5-methyl-1H-pyrazole-3-carboxylate
Molecular Weight (g/mol):
154.17
Molecular Formula:
C7H10N2O2
InChi Key:
BOTXQJAHRCGJEG-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=NNC(C)=C1
1
–
2
of
2
results
1
Ethyl 3-methyl-1H-pyrazole-5-carboxylate, 97%
CAS: 4027-57-0 Molecular Formula: C7H10N2O2 Molecular Weight (g/mol): 154.17 MDL Number: MFCD00052514 InChI Key: BOTXQJAHRCGJEG-UHFFFAOYSA-N PubChem CID: 77645 IUPAC Name: ethyl 5-methyl-1H-pyrazole-3-carboxylate SMILES: CCOC(=O)C1=NNC(C)=C1
2
Ethyl 3-methyl-1H-pyrazole-5-carboxylate, 95%, Thermo Scientific™
CAS: 4027-57-0 Molecular Formula: C7H10N2O2 Molecular Weight (g/mol): 154.17 MDL Number: MFCD00052514 InChI Key: BOTXQJAHRCGJEG-UHFFFAOYSA-N PubChem CID: 77645 IUPAC Name: ethyl 5-methyl-1H-pyrazole-3-carboxylate SMILES: CCOC(=O)C1=NNC(C)=C1