missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-methyl-1H-pyrazole-5-carboxylate, 95%, Thermo Scientific™

Product Code. 10733483
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10733483 10 g 10g
10613423 1 g 1g
10108714 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10733483 Supplier Thermo Scientific Maybridge Supplier No. SPB01217EA
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 4027-57-0
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.17
MDL Number MFCD00052514
InChI Key BOTXQJAHRCGJEG-UHFFFAOYSA-N
PubChem CID 77645
IUPAC Name ethyl 5-methyl-1H-pyrazole-3-carboxylate
SMILES CCOC(=O)C1=NNC(C)=C1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Quantity 10 g
Synonym ethyl 3-methyl-1h-pyrazole-5-carboxylate, ethyl 3-methylpyrazole-5-carboxylate, 5-methyl-1h-pyrazole-3-carboxylic acid ethyl ester, 3-methyl-1h-pyrazole-5-carboxylic acid ethyl ester, 1h-pyrazole-3-carboxylic acid, 5-methyl-, ethyl ester, ethyl3-methylpyrazole-5-carboxylate, 5-methyl-2h-pyrazole-3-carboxylic acid ethyl ester, ethyl3-methyl-1h-pyrazole-5-carboxylate, ethyl 5-methyl-2h-pyrazole-3-carboxylate, pubchem7843
Formula Weight 154.17
Percent Purity 95%
Chemical Name or Material Ethyl 3-methyl-1H-pyrazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.