accessibility menu, dialog, popup

UserName

CH3OOOHF

CAS RN 403-01-0

IUPAC Name: methyl 3-fluoro-4-hydroxybenzoate
Synonyms: methyl 3-fluoro-4-hydroxybenzoate
Molecular Weight (g/mol): 170.14
Molecular Formula: C8H7FO3
InChi Key: IYUSGKSCDUJSKS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(O)C(F)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results