accessibility menu, dialog, popup

UserName

CAS RN 403-19-0

OFNOO

CAS RN 403-19-0

IUPAC Name: 2-fluoro-4-nitrobenzen-1-olate
Synonyms: 2-fluoro-4-nitrobenzenolate
Molecular Weight (g/mol): 156.09
Molecular Formula: C6H3FNO3
InChi Key: ORPHLVJBJOCHBR-UHFFFAOYSA-M
SMILES: [O-]C1=CC=C(C=C1F)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results