accessibility menu, dialog, popup

UserName

H3CSCH3

CAS RN 40323-88-4

IUPAC Name: 2-ethyl-5-methylthiophene
Synonyms: 2-ethyl-5-methylthiophene
Molecular Weight (g/mol): 126.22
Molecular Formula: C7H10S
InChi Key: VOIVNYVBGCJFRW-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C)S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results