CAS RN 40353-34-2
CAS RN 40353-34-2
IUPAC Name:
7-nitro-1,2,3,4-tetrahydronaphthalen-1-one
Synonyms:
7-nitro-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol):
191.19
Molecular Formula:
C10H9NO3
InChi Key:
GWAQYWSNCVEJMW-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C2CCCC(=O)C2=C1
1
–
1
of
1
results
1
7-Nitro-1-tetralone, 97%
CAS: 40353-34-2 Molecular Formula: C10H9NO3 Molecular Weight (g/mol): 191.186 MDL Number: MFCD00019661 InChI Key: GWAQYWSNCVEJMW-UHFFFAOYSA-N Synonym: 7-nitro-1-tetralone,7-nitrotetralone,7-nitro-3,4-dihydronaphthalen-1 2h-one,1 2h-naphthalenone, 3,4-dihydro-7-nitro,3,4-dihydro-7-nitro-1 2h-naphthalenone,3,4-dihydro-7-nitronaphthalen-1 2h-one,wln: l66 bvt&j inw,7-nitro-2,3,4-trihydronaphthalen-1-one,1 2h-naphthalenone,4-dihydro-7-nitro,7-nitro-tetralone PubChem CID: 38445 IUPAC Name: 7-nitro-3,4-dihydro-2H-naphthalen-1-one SMILES: C1CC2=C(C=C(C=C2)[N+](=O)[O-])C(=O)C1