accessibility menu, dialog, popup

UserName

NH3F

CAS RN 404-70-6

IUPAC Name: 2-(3-fluorophenyl)ethan-1-aminium
Synonyms: 2-(3-fluorophenyl)ethanaminium
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H11FN
InChi Key: AUCVZEYHEFAWHO-UHFFFAOYSA-O
SMILES: [NH3+]CCC1=CC=CC(F)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results