missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Fluorophenyl)ethylamine, 97+%

Product Code. 15492099
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15492099 25 g 25g
15482099 5 g 5g
15402109 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15492099 Supplier Thermo Scientific Alfa Aesar Supplier No. H64762.14
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 404-70-6 | C8H11FN | 140.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 404-70-6
Molecular Formula C8H11FN
Molecular Weight (g/mol) 140.18
MDL Number MFCD00075376
InChI Key AUCVZEYHEFAWHO-UHFFFAOYSA-O
Synonym 3-fluorophenethylamine, 2-3-fluorophenyl ethylamine, 2-3-fluoro-phenyl-ethylamine, 2-3-fluorophenyl ethanamine, 2-3-fluorophenyl ethan-1-amine, benzeneethanamine, 3-fluoro, m-fluorophenethylamine, 3-flourophenethylamine, m-fluorophenylethylamine, 3-fluoro phenethylamine
PubChem CID 533928
IUPAC Name 2-(3-fluorophenyl)ethanamine
SMILES [NH3+]CCC1=CC=CC(F)=C1

Specifications

Density 1.066 g/mL
Boiling Point 87°C (15 mmHg)
Flash Point 83°C (181°F)
Refractive Index 1.509
Quantity 25 g
UN Number UN2735
Beilstein 3045789
Formula Weight 139.17
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material 2-(3-Fluorophenyl)ethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.