CAS RN 408-35-5
CAS RN 408-35-5
IUPAC Name:
sodium hexadecanoate
Synonyms:
palmitate, sodium
Molecular Weight (g/mol):
278.41
Molecular Formula:
C16H31NaO2
InChi Key:
GGXKEBACDBNFAF-UHFFFAOYSA-M
SMILES:
[Na+].CCCCCCCCCCCCCCCC([O-])=O
1
–
1
of
1
results
1
Promotions Available
Palmitic acid, sodium salt, 98%
Palmitic acid, sodium salt, 98%
CAS: 408-35-5 Molecular Formula: C16H31NaO2 Molecular Weight (g/mol): 278.4 MDL Number: MFCD00002749 InChI Key: GGXKEBACDBNFAF-UHFFFAOYSA-M Synonym: sodium palmitate,palmitic acid sodium salt,hexadecanoic acid, sodium salt,sodium hexadecanoate,palmitic acid, sodium salt,sodium pentadecanecarboxylate,unii-jq43kp6296,hsdb 759,hexadecanoic acid sodium salt,hexadecanoic acid,sodium salt 1:1 PubChem CID: 2735111 IUPAC Name: sodium;hexadecanoate SMILES: CCCCCCCCCCCCCCCC(=O)[O-].[Na+]