missing translation for 'onlineSavingsMsg'
Learn More

Palmitic acid, sodium salt, 98%

Product Code. 10393372 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10393372 5 g Glass bottle 5g
10289890 25 g Glass bottle 25g
10045403 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10393372 Supplier Thermo Scientific Acros Supplier No. 416700050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 408-35-5 | C16H31NaO2 | 278.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 408-35-5
Molecular Formula C16H31NaO2
Molecular Weight (g/mol) 278.4
MDL Number MFCD00002749
InChI Key GGXKEBACDBNFAF-UHFFFAOYSA-M
Synonym sodium palmitate, palmitic acid sodium salt, hexadecanoic acid, sodium salt, sodium hexadecanoate, palmitic acid, sodium salt, sodium pentadecanecarboxylate, unii-jq43kp6296, hsdb 759, hexadecanoic acid sodium salt, hexadecanoic acid,sodium salt 1:1
PubChem CID 2735111
IUPAC Name sodium;hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)[O-].[Na+]

Specifications

Color White
Loss on Drying 2% max. (110°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)14COONa
Quantity 5 g
Beilstein 02,III,94
Formula Weight 278.4
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Palmitic acid, sodium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.