accessibility menu, dialog, popup

UserName

OSON

CAS RN 40899-71-6

IUPAC Name: 1-(benzenesulfonyl)-1H-indole
Synonyms: 1-(benzenesulfonyl)indole
Molecular Weight (g/mol): 257.31
Molecular Formula: C14H11NO2S
InChi Key: VDWLCYCWLIKWBV-UHFFFAOYSA-N
SMILES: O=S(=O)(N1C=CC2=CC=CC=C12)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results