accessibility menu, dialog, popup

UserName

LiCH3H3CNCH3H3C

CAS RN 4111-54-0

IUPAC Name: lithium(1+) bis(propan-2-yl)azanide
Synonyms: lithium diisopropylamide
Molecular Weight (g/mol): 107.13
Molecular Formula: C6H14LiN
InChi Key: ZCSHNCUQKCANBX-UHFFFAOYSA-N
SMILES: [Li+].CC(C)[N-]C(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)